Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gstk1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89118743 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-743 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-743 Supplier Abnova Corporation Supplier No. PAB16936
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length recombinant Gstk1.

Specifications

Antigen Gstk1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant Gstk1.
Dilution Western Blot (1:2000) Immunohistochemistry (1:200) The optimal working dilution should be determined by the end user.
Formulation In serum
Gene Gstk1
Gene Alias 0610025I19Rik/AW260476
Gene Symbols Gstk1
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length mouse Gstk1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 76263
Test Specificity This antibody has been directly tested for reactivity in mouse and human. specific to the ∼25 KDa DsbA-L protein in Western blots of 3T3-L1 adipocytes and mouse white and brown adipose tissue lysate samples. It is suggested that add 1% BSA to This antibody dilution buffer.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.