Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GUCA2A Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89131675 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-131-675 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-131-675 Supplier Abnova Corporation Supplier No. PAB4993
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of GUCA2A.

Sequence: PGTCEICAYAACTGC

Specifications

Antigen GUCA2A
Applications ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GUCA2A.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal)
Gene GUCA2A
Gene Alias GUANYLIN/GUCA2/STARA
Gene Symbols GUCA2A
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human GUCA2A.
Quantity 150 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2980
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.