Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GUCY1A2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89128463 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-463 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-463 Supplier Abnova Corporation Supplier No. PAB2682
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of GUCY1A2.

Soluble guanylyl (or guanylate) cyclases are heterodimeric enzymes consisting of an alpha subunit, such as alpha-2 (GUCY1A2), and a beta subunit, typically beta-1 (GUCY1B3; MIM 139397), which are activated by nitric oxide (NO) and which catalyze conversion of GTP to 3-prime, 5-prime-cyclic GMP and pyrophosphate (Harteneck et al., 1991 [PubMed 1683630]).[supplied by OMIM

Specifications

Antigen GUCY1A2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GUCY1A2.
Dilution Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene GUCY1A2
Gene Accession No. NP_000846;P33402
Gene Alias GC-SA2/GUC1A2
Gene Symbols GUCY1A2
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human GUCY1A2.
Purification Method Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2977
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.