Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-Gustducin alpha-3 chain, Polyclonal,

Catalog No. ABN999MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN999MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN999MI Supplier MilliporeSigma™ Supplier No. ABN999
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Gustducin alpha-3 chain in Human, Rat samples, and it is validated for Immunocytochemistry, Immunohistochemistry

Gustducin alpha-3 chain is a G protein alpha subunit of the taste receptors and it plays a critical role in bitter, sweet and umami (MSG) taste transduction. Gustducin alpha-3 chain operates via the cGMP-phosphodiesterase signaling pathway and is mediated with calcium. Gustducin alpha-3 chain also functions in lumenal sugar sensors in the gut and may modulate the gut capacity to absorb sugars and thus play roles in malabsorption syndromes and diet related disorders including diabetes type II and obesity. Gustducin alpha-3 chain is localized to the cytoplasm and plasmalemmal being a G protein subunit. Gustducin alpha-3 chain is expressed in tastebuds and in the gut in enteroendocrine L cells, and surprisingly in spermatozoa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gustducin alpha-3 chain
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. A8MTJ3
Gene Symbols GNAT3
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human Gustducin alpha-3 chain near the C-terminal.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001095856
Target Species Human, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.