Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H4K8ac, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137817 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-817 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-817 Supplier Abnova Corporation Supplier No. PAB14070
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of H4K8ac.

Specifications

Antigen H4K8ac
Applications ChIP Assay, Dot Blot, ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of H4K8ac.
Dilution ELISA (1:500) Dot Blot (1:20000) Western Blot (1:250) ChIP (5 ul/ChIP) The optimal working dilution should be determined by the end user.
Formulation In serum (0.05% sodium azide)
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to region of histone H4 containing the acetylated lysine 8 (H4K8ac).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.