Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAP1 Rabbit anti-Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114354 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-354 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-354 Supplier Abnova Corporation Supplier No. PAB0702
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length recombinant Hap1.

Specifications

Antigen HAP1
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant Hap1.
Dilution Immunohistochemistry (1:500-1:3000) The optimal dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Hap1
Gene Symbols Hap1
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length rat Hap1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29430
Target Species Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.