Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HB, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89138058 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-058 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-058 Supplier Abnova Corporation Supplier No. PAB15397
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against HB.

Specifications

Antigen HB
Applications ELISA, Immunodiffusion, Radioimmune Assays (RIA), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against HB.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 20mM sodium phosphate buffer, 140mM NaCl, pH 7.3 (0.05% sodium azide)
Host Species Rabbit
Immunogen Human HB.
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Cross-reactivity to other species may exist. Antibody activity to serum proteins is not present.
Target Species Human
Content And Storage Store at 4°C on dry atmosphere.
After reconstitution with 2 mL of deionized water, store at -20°C or lower.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.