Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCRTR1 Rabbit anti-Bovine, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89117002 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-002 Supplier Abnova Corporation Supplier No. PAB14322
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of HCRTR1.

Sequence: RLYELLHGAGNHAAGILTL

Specifications

Antigen HCRTR1
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of HCRTR1.
Dilution Immunohistochemistry (1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene HCRTR1
Gene Symbols HCRTR1
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 12-33 of bovine HCRTR1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282248
Test Specificity The specificity for this antiserum has been confirmed by immunohistochemistry on rat brain and the results reflect the current literature.
Target Species Bovine, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.