Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDLBP Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89121772 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-772 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-772 Supplier Abnova Corporation Supplier No. PAB20269
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant HDLBP.

High density lipoprotein-binding protein, also known as vigilin, is a 110kDa protein that specifically binds HDL molecules and may function in the removal of excess cellular cholesterol.[supplied by OMIM

Sequence: DMNQFGEGEQAKICLEIMQRTGAHLELSLAKDQGLSIMVSGKLDAVMKARKDIVARLQTQASATVAIPKEHHRFVIGKNGEKLQDLELKTATKIQIPRPDDPSNQIKITGTKEGIEKARHEVLLISAEQDKRAVE

Specifications

Antigen HDLBP
Applications Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant HDLBP.
Dilution Immunohistochemistry (1:200-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene HDLBP
Gene Alias FLJ16432/HBP/PRO2900/VGL
Gene Symbols HDLBP
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human HDLBP.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3069
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.