Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HA Tag Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB0601
This item is not returnable.
View return policy
Description
HA Tag Monoclonal specifically detects HA Tag in Multi-Species samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Immunocytochemistry.Specifications
HA Tag | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with Sodium Azide | |
HA, HA epitope tag, Hemagglutinin | |
HA Tag peptide Accession # ABB51961 | |
50 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Western Blot, Immunoprecipitation, Immunocytochemistry | |
1049F | |
Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunoprecipitation 1-4 ug/100 ug cell lysate, Immunocytochemistry 8-25 ug/mL | |
ABB51961 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Detects N-terminal and C-terminal HA-tagged proteins in Western blot. | |
Multi-species | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction