Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hepatitis B virus core antigen, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137943 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-943 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-943 Supplier Abnova Corporation Supplier No. PAB14506
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against hepatitis B virus core antigen.

Specifications

Antigen hepatitis B virus core antigen
Applications Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against hepatitis B virus core antigen.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:400) The optimal working dilution should be determined by the end user.
Formulation In 10mM phosphate buffer, 150mM NaCl, pH 7.4 (0.05% sodium azide)
Host Species Rabbit
Immunogen Native purified virus hepatitis B virus core antigen.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Hepatitis B virus
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.