Learn More
hepatitis C virus core Polyprotein Rabbit anti-Viruses, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Antigen | hepatitis C virus core Polyprotein |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic peptide of hepatitis C virus core Polyprotein. |
| Dilution | ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulation | In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to N-terminus of HCV core Polyprotein. |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.