Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hepatitis E virus (HEV), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89136921 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-921 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-921 Supplier Abnova Corporation Supplier No. PAB0920

Rabbit polyclonal antibody raised against synthetic peptide of hepatitis E virus (HEV).

Sequence: CPECRTLGLQG

Specifications

Antigen hepatitis E virus (HEV)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS (0.02% sodium azide)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 627-637 of HEV capsid protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.