Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HGFAC, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89118072 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-072 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-072 Supplier Abnova Corporation Supplier No. PAB16182
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant HGFAC.

The protein encoded by this gene belongs to peptidase family S1. It is first synthesized as an inactive single-chain precursor before being activated to a heterodimeric form by endoproteolytic processing. It acts as serine protease that converts hepatocyte growth factor to the active form. [provided by RefSeq

Specifications

Antigen HGFAC
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant HGFAC.
Dilution ELISA (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS.
Gene HGFAC
Gene Alias HGFA/MGC138395/MGC138397
Gene Symbols HGFAC
Host Species Rabbit
Immunogen Recombinant protein corresponding to human HGFAC.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3083
Target Species Human
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.