Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HICE1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121439 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-439 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-439 Supplier Abnova Corporation Supplier No. PAB19929
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length recombinant HICE1.

Specifications

Antigen HICE1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant HICE1.
Dilution ELISA (1:250000) Western Blot (1:10000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene HICE1
Gene Alias MGC20533/NY-SAR-48
Gene Symbols HICE1
Host Species Rabbit
Immunogen Recombinant GST fusion protein corresponding to full length human HICE1.
Purification Method Immunoaffinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93323
Test Specificity This antibody reacts with endogenous Hice1 protein. A BLAST analysis was used to suggest reactivity with Hice1 from human based on a 100% homology with the immunizing sequence. Expect reactivity with Hice1 from chimpanzee, Sumatran orangutan based on a 90% homology with the immunizing sequence.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.