Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF1A Rabbit anti-Human, Mouse, Primates, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89115636 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-636 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-636 Supplier Abnova Corporation Supplier No. PAB12138
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Hif1a.

Specifications

Antigen HIF1A
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Hif1a.
Dilution Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Frozen sections) (5-10 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) Western Blot (1:500-1:2000) The optimal working dil
Formulation In PBS (0.05% sodium azide)
Gene Hif1a
Gene Alias AA959795/HIF-1alpha/HIF1alpha/MOP1
Gene Symbols Hif1a
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 530-825 of mouse Hif1a.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15251
Test Specificity This antibody is specific to mouse HIF-1 alpha.
Target Species Human, Mouse, Primate, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.