Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF1A Rabbit anti-Human, Mouse, Primates, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89115791 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-791 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-791 Supplier Abnova Corporation Supplier No. PAB12418
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant HIF1A.

Hypoxia-inducible factor-1 (HIF1) is a transcription factor found in mammalian cells cultured under reduced oxygen tension that plays an essential role in cellular and systemic homeostatic responses to hypoxia. HIF1 is a heterodimer composed of an alpha subunit and a beta subunit. The beta subunit has been identified as the aryl hydrocarbon receptor nuclear translocator (ARNT). This gene encodes the alpha subunit of HIF-1. Overexpression of a natural antisense transcript (aHIF) of this gene has been shown to be associated with nonpapillary renal carcinomas. Two alternative transcripts encoding different isoforms have been identified. [provided by RefSeq

Specifications

Antigen HIF1A
Applications ChIP Assay, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant HIF1A.
Dilution Western Blot (1:500-1:1000) Immunohistochemistry (1:100) ChIP (25 ul/1.5 x 106 cells) The optimal working dilution should be determined by the end user.
Formulation In antiserum (0.05% sodium azide)
Gene HIF1A
Gene Alias HIF-1alpha/HIF1/HIF1-ALPHA/MOP1/PASD8/bHLHe78
Gene Symbols HIF1A
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 432-528 of human HIF1A.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3091
Test Specificity This antibody is specific to HIF-1 alpha.
Target Species Human, Mouse, Primate, Rat
Content And Storage Store at 4°C for short term storage. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.