Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF3A Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114742 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-742 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-742 Supplier Abnova Corporation Supplier No. PAB10271
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Hif3a.

Specifications

Antigen HIF3A
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Hif3a.
Dilution ELISA (1:14000-1:80000) Western Blot (1:10500-1:8000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene Hif3a
Gene Alias Ipas/MOP7
Gene Symbols Hif3a
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 581-592 of mouse Hif3a.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53417
Test Specificity This affinity-purified antibody is directed against mouse Hif3alpha protein. No reactivity is expected against other forms of Hif proteins.
Target Species Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.