Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIGD2A, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HIGD2A.
Supplier: Abnova Corporation PAB25761
Specifications
HIGD2A | |
Polyclonal | |
Unconjugated | |
Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
HIGD2A | |
HIGD2A | |
A synthetic peptide corresponding to 16 amino acids at C-terminus of human HIGD2A. | |
100 μg | |
Primary | |
At least two isoforms of HIG2 are known to exist; this antibody will detect both isoforms. HIG2 antibody is predicted to not cross-react with HIG1. | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
ELISA, Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of HIGD2A. | |
In PBS (0.02% sodium azide) | |
MGC2198 | |
Rabbit | |
Peptide affinity purification | |
RUO | |
192286 | |
Human, Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction