Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

histone H3 (acetylated lysines 9 and 14), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89136889 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-889 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-889 Supplier Abnova Corporation Supplier No. PAB0652
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of histone H3 (acetylated lysines 9 and 14).

Specifications

Antigen histone H3 (acetylated lysines 9 and 14)
Applications ChIP Assay, Dot Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of histone H3 (acetylated lysines 9 and 14).
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.1% BSA, 0.01% thimerosal)
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) containing the acetylated lysines 9 and 14 (or [K9/14ac]) of human histone H3.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.