Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3 (K27ac), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121293 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-293 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-293 Supplier Abnova Corporation Supplier No. PAB19766
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Histone H3 (K27ac).

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is located separately from the other H3 genes that are in the histone gene cluster on chromosome 6p22-p21.3. [provided by RefSeq

Specifications

Antigen Histone H3 (K27ac)
Applications ChIP Assay, Dot Blot, ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Histone H3 (K27ac).
Dilution ELISA (1:100) Western Blot (1:1000) ChIP/ChIP-seq (1-2 ug/ChIP) Dot Blot (1:25000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide, 0.05% Proclin 300)
Gene HIST3H3
Gene Alias H3.4/H3/g/H3FT/H3t/MGC126886/MGC126888
Gene Symbols HIST3H3
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to Histone H3, acetylated at lysine 27.
Purification Method Affinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8290
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.