Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3 (K4me2), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Histone H3 (K4me2).
Supplier: Abnova Corporation PAB15453
Specifications
| Histone H3 (K4me2) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Histone H3 (K4me2). | |
| In PBS, pH 7.2 (0.09% sodium azide) | |
| Lysine4 dimethylated peptide of Histone H3. | |
| RUO | |
| This antibody is expected to react with Histone H3 from yeast to mammals. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ChIP Assay, Dot Blot, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Yeast | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction