Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV-1 Gag p55 protein, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant HIV-1 Gag p55 protein.
Supplier: Abnova Corporation PAB1181
Specifications
HIV-1 Gag p55 protein | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant HIV-1 Gag p55 protein. | |
In buffer containing 0.09% sodium azide | |
Recombinant protein corresponding to human immunodeficiency virus type 1 Gag p55 protein. | |
RUO | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Rabbit | |
50 μL | |
Primary | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction