Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV-1 Nef protein, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137480 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-480 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-480 Supplier Abnova Corporation Supplier No. PAB1182

Rabbit polyclonal antibody raised against recombinant HIV-1 Nef protein.

Specifications

Antigen HIV-1 Nef protein
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant HIV-1 Nef protein.
Dilution Dot Blot (1:3000) ELISA Immunoprecipitation Western Blot (1:1000 - 1:3000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Host Species Rabbit
Immunogen Recombinant protein corresponding to human immunodeficiency virus type 1 Nef protein.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at -20°C.
Avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.