Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV-1 p24, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89138421 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-421 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-421 Supplier Abnova Corporation Supplier No. PAB27384
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length recombinant HIV-1 p24.

Specifications

Antigen HIV-1 p24
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant HIV-1 p24.
Dilution Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Host Species Rabbit
Immunogen Recombinant protein corresponding to human immunodeficiency virus type 1 p24.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity In HIV-1 samples, p24 antibody detects a ∼24 kDa, a ∼41 kDa, and a ∼55 kDa protein, corresponding to HIV-1 p24 and its precursors p41 and p55, respectively.
Target Species Virus
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.