Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV-1 reverse transcriptase, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant HIV-1 reverse transcriptase.
Supplier: Abnova Corporation PAB1174
Specifications
HIV-1 reverse transcriptase | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant HIV-1 reverse transcriptase. | |
In buffer containing 0.09% sodium azide | |
Recombinant protein corresponding to human immunodeficiency virus type 1 reverse transcriptase. | |
RUO | |
Store at 4°C. |
ELISA, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Rabbit | |
50 μL | |
Primary | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction