Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIVEP3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89123625 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-625 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-625 Supplier Abnova Corporation Supplier No. PAB21918
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant HIVEP3.

Members of the ZAS family, such as ZAS3 (HIVEP3), are large proteins that contain a ZAS domain, a modular protein structure consisting of a pair of C2H2 zinc fingers with an acidic-rich region and a serine/threonine -rich sequence. These proteins bind specific DNA sequences, including the kappa-B motif (GGGACTTTCC), in the promoters and enhancer regions of several genes and viruses, including human immunodeficiency virus (HIV). ZAS genes span more than 150kb and contain at least 10 exons, one of which is longer than 5.5kb (Allen and Wu, 2004).[supplied by OMIM

Sequence: SYSFDDHITDSEALSRSSHVFTSHPRMLKRQPAIELPLGGEYSSEEPGPSSKDTASKPSDEVEPKESELTKKTKKGLKTKGVIYECNICGARYKKRDNYEAHKKYYCSELQIAKPISAGTHTSPEAEKSQIEHEPWSQMMHYKLGTTL

Specifications

Antigen HIVEP3
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant HIVEP3.
Dilution Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene HIVEP3
Gene Alias FLJ16752/KBP-1/KBP1/KIAA1555/KRC/SHN3/Schnurri-3/ZAS3/ZNF40C
Gene Symbols HIVEP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human HIVEP3.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59269
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.