Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-HNF-6, Polyclonal,

Catalog No. ABN1022MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1022MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1022MI Supplier MilliporeSigma™ Supplier No. ABN1022
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects HNF-6 in Human samples, and it is validated for Western Blotting

Hepatocyte nuclear factor 6 (UniProt Q9UBC0; also known as HNF-6, One cut domain family member 1, One cut homeobox 1) is a nucleus protein encoded by the ONECUT1 (also known as HNF6, HNF6A) gene (Entrez Gene ID 3175) in human. HNF-6 is a member of the Cut homeobox family of transcription factors. HNF-6 is highly expressed in the liver, where it regulates the expression of targets genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HNF-6
Applications Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9UBC0
Gene Symbols ONECUT1; HNF6; HNF6A
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human HNF-6 near the C-terminus.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_004489
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.