Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNRNPUL2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89119412 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-412 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-412 Supplier Abnova Corporation Supplier No. PAB17746
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of HNRNPUL2.

Specifications

Antigen HNRNPUL2
Applications ELISA, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of HNRNPUL2.
Dilution Immunohistochemistry (1:50-1:100) ELISA (1:40000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Gene HNRNPUL2
Gene Alias DKFZp762N1910/HNRPUL2
Gene Symbols HNRNPUL2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal of human HNRNPUL2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221092
Test Specificity This antibody detects endogenous levels of total HNRNPUL2 protein
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.