Learn More
HOXA1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HOXA1.
Supplier: Abnova Corporation PAB19786
Description
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. The encoded protein may be involved in the placement of hindbrain segments in the proper location along the anterior-posterior axis during development. Two transcript variants encoding two different isoforms have been found for this gene, with only one of the isoforms containing the homeodomain region. [provided by RefSeq
Specifications
HOXA1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of HOXA1. | |
In serum (0.5% BSA, 0.02% sodium azide) | |
BSAS/HOX1/HOX1F/MGC45232 | |
Rabbit | |
Peptide affinity purification | |
RUO | |
3198 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:20000-1:80000) Western Blot (1:200-1000) The optimal working dilution should be determined by the end user. | |
HOXA1 | |
HOXA1 | |
A synthetic peptide corresponding to 15 amino acids at internal region of human HOXA1. | |
100 μg | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only