Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-HOXA10, Polyclonal, Abnova™

Catalog No. 89117167 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-167 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-167 Supplier Abnova Corporation Supplier No. PAB14592

Specifications

Antigen HOXA10
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS (0.2% gelatin, 0.05% sodium azide)
Gene Symbols HOXA10
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 263-277 of human HOXA10.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3206
Target Species Human, Monkey
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.