Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-HOXA5, Polyclonal, Abnova™

Catalog No. 89117213 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-213 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-213 Supplier Abnova Corporation Supplier No. PAB14649

Specifications

Antigen HOXA5
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation In serum (50% glycerol, 0.025% sodium azide)
Gene Symbols HOXA5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 142-161 of human HOXA5.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 3202
Target Species Bovine, Canine, Chicken, Chimpanzee, Human, Mouse
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.