Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HP Rabbit anti-Bovine, Polyclonal Antibody, Abnova™

Catalog No. 89121426 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-426 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-426 Supplier Abnova Corporation Supplier No. PAB19915
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native HP.

Specifications

Antigen HP
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native HP.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (0.02% sodium azide, 50% glycerol (v/v))
Gene HP
Gene Alias MGC133752
Gene Symbols HP
Host Species Rabbit
Immunogen Native purified HP from bovine plasma.
Purification Method Affinity purification
Quantity 150 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 280692
Target Species Bovine
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.