Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRAS/NRAS/KRAS, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115467 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-467 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-467 Supplier Abnova Corporation Supplier No. PAB11953

Rabbit polyclonal antibody raised against synthetic peptide of HRAS/NRAS/KRAS.

Sequence: EYDPTIEDSYRKQ

Specifications

Antigen HRAS/NRAS/KRAS
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of HRAS/NRAS/KRAS.
Dilution Western Blot (1-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In BBS, pH 8.2
Gene Alias C-BAS/HAS/C-H-RAS/C-HA-RAS1/CTLO/H-RASIDX/HAMSV/HRAS1/K-RAS/N-RAS/RASH1
Gene Symbols HRAS
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 31-43 of human HRAS/NRAS/KRAS.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3265/4893/3845
Test Specificity This antibody reacts with recombinant GST Ras and GST Ras fusion proteins as well as with 21 and 23 KDa bands. The pp23 band is believed to represent the phosphorylated form of p21. It does not cross-react with members of the Rac, Rho, or Rab families of Ras-related small G-proteins.
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.