Learn More
HRH3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HRH3.
Supplier: Abnova Corporation PAB12272
Description
Histamine is a ubiquitous messenger molecule released from mast cells, enterochromaffin-like cells, and neurons. Its various actions are mediated by histamine receptors H1, H2, H3 and H4. This gene encodes one of the histamine receptors (H3) which belongs to the family 1 of G protein-coupled receptors. It is an integral membrane protein and can regulate neurotransmitter release. This receptor can also increase voltage-dependent calcium current in smooth muscles and innervates the blood vessels and the heart in cardiovascular system. [provided by RefSeq
Specifications
HRH3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of HRH3. | |
Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
GPCR97/HH3R | |
Rabbit | |
Affinity purification | |
RUO | |
11255 | |
Human, Mouse, Rat | |
Lyophilized |
Western Blot | |
Unconjugated | |
Western Blot (0.75 ug/mL) The optimal working dilution should be determined by the end user. | |
HRH3 | |
HRH3 | |
A synthetic peptide corresponding to amino acids at C-terminus of human HRH3. | |
100 μg | |
Primary | |
Different to the related rat sequence by two amino acides. | |
Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only