Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-HSD17B7, Polyclonal,

Catalog No. ABS1009MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1009MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1009MI Supplier MilliporeSigma™ Supplier No. ABS1009
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects HSD17B7 in Bovine, Hamster, Human, Mouse, Rat samples, and it is validated for Immunocytochemistry, Immunohistochemistry, Western Blotting

3-keto-steroid reductase (UniProt Q62904; EC 1.1.1.270, EC 1.1.1.62; also known as PRAP, 17-beta-HSD 7, PRL receptor-associated protein, Estradiol 17-beta-dehydrogenase 7, 17-beta-hydroxysteroid dehydrogenase 7, Hydroxysteroid dehydrogenase 17 beta, type 7) is encoded by the Hsd17b7 gene in rat (Entrez Gene ID 29540). Hsd17b7 protein is most abundantly expressed in ovaries of pregnant animals and functions both as a 17-beta-hydroxysteroid dehydrogenase (EC 1.1.1.62) in the biosynthesis of sex steroids, as well as as a 3-ketosteroid reductase (EC 1.1.1.270) in the biosynthesis of cholesterol. Anti-HSD17B7, is a rabbit polyclonal antiserum raised against native rat protein, and shows reactivity toward rat, human, mouse, hamster, and bovine species (PMID 1425419).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B7
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Accession No. Q62904
Gene Symbols Hsd17b7
Host Species Rabbit
Immunogen Purified protein corresponding to rat HSD17B7.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_058931
Target Species Bovine, Hamster, Human, Mouse, Rat
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.