Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSF1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89133925 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-133-925 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-133-925 Supplier Abnova Corporation Supplier No. PAB8772

Rabbit polyclonal antibody raised against synthetic peptide of Hsf1.

Specifications

Antigen HSF1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description heat shock factor 1
Formulation In PBS (30% glycerol, 0.5% BSA, 0.01% thimerosal)
Gene Hsf1
Gene Alias AA960185
Gene Symbols Hsf1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues surrounding amino acids 515 of mouse Hsf1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15499
Test Specificity This antibody recognizes 100-120kDa band, corresponding to glycosylated Hsf1.
Target Species Human, Mouse
Content And Storage Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.