Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137940 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-940 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-940 Supplier Abnova Corporation Supplier No. PAB14473
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against Hsp90.

Specifications

Antigen HSP90
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Hsp90.
Dilution Western Blot (1:20000-1:40000) The optimal working dilution should be determined by the end user.
Formulation In serum
Host Species Rabbit
Immunogen Full length Hsp90 protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes eukaryotic Hsp90alpha/beta.
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.