Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90AA1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89116748 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-748 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-748 Supplier Abnova Corporation Supplier No. PAB13569
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Hsp90aa1.

Sequence: PEETQTQDQPM-Cys

Specifications

Antigen HSP90AA1
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Hsp90aa1.
Dilution Western blot (5-10 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) Immunoprecipitation (10 ug/mg protein lysate) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (0.09% sodium azide)
Gene Hsp90aa1
Gene Alias 86kDa/89kDa/AL024080/AL024147/Hsp86-1/Hsp89/Hsp90/Hspca/hsp4
Gene Symbols Hsp90aa1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 2-12 of mouse Hsp90aa1.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15519
Test Specificity This antibody recognizes a protein of 86 KDa identified as heat shock protein 90alpha. Anti-HSP90alpha weakly cross-reacts with HSP90beta.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.