Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90AB1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89120431 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-431 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-431 Supplier Abnova Corporation Supplier No. PAB18870
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of HSP90AB1.

HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. There are 2 major cytosolic HSP90 proteins, HSP90AA1 (MIM 140571), an inducible form, and HSP90AB1, a constitutive form. Other HSP90 proteins are found in endoplasmic reticulum (HSP90B1; MIM 191175) and mitochondria (TRAP1; MIM 606219) (Chen et al., 2005 [PubMed 16269234]).[supplied by OMIM

Specifications

Antigen HSP90AB1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of HSP90AB1.
Dilution ELISA (1:80000) Western Blot (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene HSP90AB1
Gene Alias D6S182/FLJ26984/HSP90-BETA/HSP90B/HSPC2/HSPCB
Gene Symbols HSP90AB1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 14 amino acids at N-terminus of human HSP90AB1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3326
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.