Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90B1, Rabbit, ATTO390, Polyclonal Antibody, Abnova™

Catalog No. 89130474 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-474 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-474 Supplier Abnova Corporation Supplier No. PAB28947
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Hsp90b1.

Specifications

Antigen HSP90B1
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Atto 390
Description Rabbit polyclonal antibody raised against synthetic peptide of Hsp90b1.
Dilution Immunocytochemistry Immunohistochemistry Immunoprecipitation (1:80) Western blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Hsp90b1
Gene Alias ERp99/GRP94/TA-3/Targ2/Tra-1/Tra1/gp96
Gene Symbols Hsp90b1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids at C-terminus of mouse Hsp90b1.
Purification Method Peptide Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22027
Test Specificity Detects ∼94 kDa.
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.