Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90B1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89116749 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-749 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-749 Supplier Abnova Corporation Supplier No. PAB13570

Rabbit polyclonal antibody raised against synthetic peptide of Hsp90b1.

Sequence: PEEVHHGEEEVE-Cys

Specifications

Antigen HSP90B1
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Hsp90b1.
Dilution Western blot (5-10 ug/mL) Immunoprecipitation (10 ug/mg protein lysate) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (0.09% sodium azide)
Gene Hsp90b1
Gene Alias ERp99/GRP94/TA-3/Targ2/Tra-1/Tra1/gp96
Gene Symbols Hsp90b1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 2-13 of mouse Hsp90b1.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22027
Test Specificity This antibody recognizes a protein of 84 KDa identified as heat shock protein 90beta. Anti-HSP90beta does not cross-react with HSP90alpha.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.