Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hspa4, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117765 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-765 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-765 Supplier Abnova Corporation Supplier No. PAB15700
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Hspa4.

Sequence: LNLQNKQSLTVDPVVKTKEIEAKIKELTSICSPIISKPKPKVEPPKEEPKHAEQNGPVDGQGDNPGSQAAEHGADTAVPSDGDKKLPEMDID

Specifications

Antigen Hspa4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Hspa4.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.02% sodium azide)
Gene Hspa4
Gene Alias 70kDa/AI317151/APG-2/Hsp110/Hsp70RY/KIAA4025/mKIAA4025
Gene Symbols Hspa4
Host Species Rabbit
Immunogen Recombinant GST fusion protein corresponding to 92 mouse Hspa4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15525
Test Specificity Specific to recombinant protein GX0631. This antibody detects endogenous mHSPA4 protein in several cell types. It also recognizes human HSPA4 protein.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.