Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPE1 Rabbit anti-Bovine, Dog, Frog, Guinea pig, Human, Mouse, Pig, Rabbit, Rat, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89117617 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-617 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-617 Supplier Abnova Corporation Supplier No. PAB15431
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of HSPE1.

This gene encodes a major heat shock protein which functions as a chaperonin. Its structure consists of a heptameric ring which binds to another heat shock protein in order to form a symmetric, functional heterodimer which enhances protein folding in an ATP-dependent manner. This gene and its co-chaperonin, HSPD1, are arranged in a head-to-head orientation on chromosome 2. [provided by RefSeq

Specifications

Antigen HSPE1
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of HSPE1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.3 (0.02% sodium azide)
Gene HSPE1
Gene Alias CPN10/GROES/HSP10
Gene Symbols HSPE1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 91-101 of human HSPE1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3336
Test Specificity Detects ∼ 10KDa.
Target Species Bovine, Canine, Frog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sheep
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.