Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTR1F Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™

Catalog No. 89127148 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-148 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-148 Supplier Abnova Corporation Supplier No. PAB25534
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of HTR1F.

Specifications

Antigen HTR1F
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of HTR1F.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (6-9 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene HTR1F
Gene Alias 5-HT1F/5HT6/HTR1EL/MR77
Gene Symbols HTR1F
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 19 amino acids at 3rd cytoplasmic domain of human HTR1F.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3355
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins, except TPRXL (47%).
Target Species Human, Monkey
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.