Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTR2A Rabbit anti-Horse, Human, Pig, Polyclonal Antibody, Abnova™

Catalog No. 89127156 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-156 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-156 Supplier Abnova Corporation Supplier No. PAB25542
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of HTR2A.

Specifications

Antigen HTR2A
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of HTR2A.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10-12.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene HTR2A
Gene Alias 5-HT2A/HTR2
Gene Symbols HTR2A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 17 amino acids at N-terminus of human HTR2A.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3356
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Target Species Equine, Human, Pig
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.