Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aspergillus, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137723 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-723 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-723 Supplier Abnova Corporation Supplier No. PAB13893
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against Aspergillus.

Specifications

Antigen Aspergillus
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Aspergillus.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Rabbit
Immunogen Soluble extract from Aspergillus fumigatus, Aspergillus flavus, Aspergillus niger and Aspergillus terreus.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reactive with soluble proteins from common Aspergillus species. Antiserum is not absorbed and may react with related microorganisms.
Target Species Bacteria, Fungi
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.