Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clostridium species, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Clostridium species.
Supplier: Abnova Corporation PAB13918
Specifications
| Clostridium species | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against Clostridium species. | |
| In 10mM PBS, pH 7.2 (0.09% sodium azide) | |
| Native Clostridium perfingens, Clostridium sordellii, Clostridium novyi, Clostridium septicum and Clostridium chauvoei. | |
| RUO | |
| Reactive with proteins from common Clostridia species. Antiserum is not absorbed and may react with related microorganisms. Reactive (IFA) with Cl. perfingens, Cl. sordellii, Cl. novyi, Cl. septicum, Cl. chauvoei. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 1 mL | |
| Primary | |
| Clostridium species | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction