Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Escherichia coli (Alkaline Phosphatase), Polyclonal, Abnova™

Catalog No. 89137808 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-808 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-808 Supplier Abnova Corporation Supplier No. PAB14039

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Antigen Escherichia coli (Alkaline Phosphatase)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Host Species Rabbit
Immunogen Alkaline Phosphatase from Escherichia coli.
Quantity 2 mL
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.