Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Escherichia coli Maltose Binding Protein Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137680 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-680 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-680 Supplier Abnova Corporation Supplier No. PAB13758

Rabbit polyclonal antibody raised against recombinant Maltose Binding Protein.

Specifications

Antigen Escherichia coli Maltose Binding Protein
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant Maltose Binding Protein.
Dilution ELISA (Detection: 1:10000 Capture: 1:200-1:500) Western Blot (1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Host Species Rabbit
Immunogen Recombinant protein corresponding to Escherichia coli Maltose Binding Protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.